missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11066-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3′ of the DNA lesion while the ERCC1 / XPF complex makes the 5′ incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.Spécification
ABCB9 | |
Polyclonal | |
Unconjugated | |
ABCB9 | |
ABC transporter 9 protein, ABCB9, EST122234, hABCB9, KIAA1520, TAP like protein, TAPL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23457, 56325 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9JJ59, Q9NP78 | |
Abcb9 | |
ABCB9 Fusion Protein Ag1542 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |