Learn More
Acide 4-méthoxy-3-méthylbenzèneboronique, 98+ %, Thermo Scientific Chemicals
25.40€ - 87.60€
Identifiants chimiques
CAS | 175883-62-2 |
---|---|
Formule moléculaire | C8H11BO3 |
Poids moléculaire (g/mol) | 165.98 |
Numéro MDL | MFCD02179464 |
Clé InChI | PXVDQGVAZBTFIB-UHFFFAOYSA-N |
Synonyme | 4-methoxy-3-methylbenzeneboronic acid, 4-methoxy-3-methylphenyl boronic acid, 4-methoxy-3-methylphenyl boranediol, 3-methyl-4-methoxyphenylboronic acid, boronic acid, 4-methoxy-3-methylphenyl, boronic acid,b-4-methoxy-3-methylphenyl, b-4-methoxy-3-methylphenyl boronic acid, akos brn-0220, 4-methoxy-3-methyl-phenyl boronic acid, timtec-bb sbb005903 |
CID PubChem | 2773487 |
Nom IUPAC | Acide (4-méthoxy-3-méthylphényl) boronique |
SMILES | COC1=CC=C(C=C1C)B(O)O |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
11359795
|
Thermo Scientific Alfa Aesar
L19821.03 |
1 g |
25.40€
1g |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
11369795
|
Thermo Scientific Alfa Aesar
L19821.06 |
5 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
175883-62-2 | |
165.98 | |
PXVDQGVAZBTFIB-UHFFFAOYSA-N | |
2773487 | |
COC1=CC=C(C=C1C)B(O)O |
C8H11BO3 | |
MFCD02179464 | |
4-methoxy-3-methylbenzeneboronic acid, 4-methoxy-3-methylphenyl boronic acid, 4-methoxy-3-methylphenyl boranediol, 3-methyl-4-methoxyphenylboronic acid, boronic acid, 4-methoxy-3-methylphenyl, boronic acid,b-4-methoxy-3-methylphenyl, b-4-methoxy-3-methylphenyl boronic acid, akos brn-0220, 4-methoxy-3-methyl-phenyl boronic acid, timtec-bb sbb005903 | |
Acide (4-méthoxy-3-méthylphényl) boronique |
Spécification
4-Methoxy-3-methylbenzeneboronic acid | |
∼215°C | |
MFCD02179464 | |
4-methoxy-3-methylbenzeneboronic acid, 4-methoxy-3-methylphenyl boronic acid, 4-methoxy-3-methylphenyl boranediol, 3-methyl-4-methoxyphenylboronic acid, boronic acid, 4-methoxy-3-methylphenyl, boronic acid,b-4-methoxy-3-methylphenyl, b-4-methoxy-3-methylphenyl boronic acid, akos brn-0220, 4-methoxy-3-methyl-phenyl boronic acid, timtec-bb sbb005903 | |
COC1=CC=C(C=C1C)B(O)O | |
165.98 | |
165.99 |
175883-62-2 | |
C8H11BO3 | |
7475212 | |
PXVDQGVAZBTFIB-UHFFFAOYSA-N | |
Acide (4-méthoxy-3-méthylphényl) boronique | |
2773487 | |
≥98% |
Sécurité et traitement
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only