Learn More
4-chloro-2,6-diméthylphénol, 98 %, Thermo Scientific Chemicals
Code produit. 11460234
Click to view available options
Quantité:
10 g
50 g
250 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 1123-63-3
Formule moléculaire C8H9ClO
Poids moléculaire (g/mol) 156.609
Numéro MDL MFCD00020138
Clé InChI VWYKSJIPZHRLNO-UHFFFAOYSA-N
CID PubChem 70747
Nom IUPAC 4-chloro-2,6-diméthylphénol
SMILES CC1=CC(=CC(=C1O)C)Cl
Spécification
Nom chimique ou matériau 4-Chloro-2,6-dimethylphenol
Point de fusion 80°C to 83°C
Quantité 250 g
Beilstein 1908282
Synonyme 4-chloro-2,6-xylenol, phenol, 4-chloro-2,6-dimethyl, 2,6-dimethyl-4-chlorophenol, 4-chloro-2,6-dimethyl-phenol, 2,6-xylenol, 4-chloro, pubchem2657, acmc-1c4bc, phenol,4-chloro-2,6-dimethyl, 4-chloro-2,6-xylenol oh=1, 4-chloro-2,6-dimethylphenol
Poids de la formule 156.61
Pourcentage de pureté 98%