Learn More
4’-bromonanophénone, 98 %, Thermo Scientific™
Code produit. 11475650
Click to view available options
Quantité:
5 g
25 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 126334-26-7
Formule moléculaire C15H21BrO
Poids moléculaire (g/mol) 297.236
Numéro MDL MFCD09953463
Clé InChI JIVVSHXGCFTTTG-UHFFFAOYSA-N
Synonyme 1-4-bromophenyl nonan-1-one, 4'-bromononanophenone, 1-nonanone,1-4-bromophenyl, acmc-20e7mb, 4-nonanoyl bromobenzene, 4-bromo-alpha-octylbenzaldehyde
CID PubChem 23117403
Nom IUPAC 1-(4-bromophényl)nonan-1-one
SMILES CCCCCCCCC(=O)C1=CC=C(C=C1)Br
Spécification
Nom chimique ou matériau 4'-Bromononanophenone
Point de fusion 65°C to 69°C
Point d’ébullition 138°C to 140°C (0.1 mmHg)
Quantité 5 g
Poids de la formule 297.24
Pourcentage de pureté 98%