Learn More
4-(3-Pipéridinyloxy)benzonitrile, 97 %, Thermo Scientific™
64.20€ - 244.00€
Identifiants chimiques
CAS | 902837-34-7 |
---|---|
Formule moléculaire | C12H14N2O |
Poids moléculaire (g/mol) | 202.257 |
Numéro MDL | MFCD08061105 |
Clé InChI | KMBKWCFPFFJUPE-UHFFFAOYSA-N |
Synonyme | 4-3-piperidinyloxy benzonitrile, 4-piperidin-3-yloxy benzonitrile, 3-4-cyanophenoxy piperidine hcl, 4-piperidin-3-yl oxy benzonitrile |
CID PubChem | 44721293 |
Nom IUPAC | 4-pipéridine-3-yloxybenzonitrile |
SMILES | C1CC(CNC1)OC2=CC=C(C=C2)C#N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
902837-34-7 | |
202.257 | |
KMBKWCFPFFJUPE-UHFFFAOYSA-N | |
44721293 | |
C1CC(CNC1)OC2=CC=C(C=C2)C#N |
C12H14N2O | |
MFCD08061105 | |
4-3-piperidinyloxy benzonitrile, 4-piperidin-3-yloxy benzonitrile, 3-4-cyanophenoxy piperidine hcl, 4-piperidin-3-yl oxy benzonitrile | |
4-pipéridine-3-yloxybenzonitrile |
Spécification
4-(3-Piperidinyloxy)benzonitrile | |
C12H14N2O | |
4-3-piperidinyloxy benzonitrile, 4-piperidin-3-yloxy benzonitrile, 3-4-cyanophenoxy piperidine hcl, 4-piperidin-3-yl oxy benzonitrile | |
C1CC(CNC1)OC2=CC=C(C=C2)C#N | |
202.257 | |
202.26 |
902837-34-7 | |
MFCD08061105 | |
KMBKWCFPFFJUPE-UHFFFAOYSA-N | |
4-pipéridine-3-yloxybenzonitrile | |
44721293 | |
97% |
Sécurité et traitement
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Température ambiante