Learn More
3-Acétyl-2,5-dichlorothiophène, 98 %, Thermo Scientific Chemicals
Code produit. 11411548
Click to view available options
Quantité:
5 g
25 g
100 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 36157-40-1
Formule moléculaire C6H4Cl2OS
Poids moléculaire (g/mol) 195.057
Numéro MDL MFCD00014522
Clé InChI GYFDNIRENHZKGR-UHFFFAOYSA-N
Synonyme 3-acetyl-2,5-dichlorothiophene, 1-2,5-dichlorothiophen-3-yl ethanone, 1-2,5-dichloro-3-thienyl ethan-1-one, 1-2,5-dichlorothiophen-3-yl ethan-1-one, 1-2,5-dichloro-3-thienyl ethanone, 2,5-dichloro-3-thienyl methyl ketone, ethanone, 1-2,5-dichloro-3-thienyl, 1-2,5-dichloro-thiophen-3-yl-ethanone, 1-2,5-dichlorothien-3-yl ethan-1-one, zlchem 481
CID PubChem 118920
Nom IUPAC 1-(2,5-dichlorothiophène-3-yl)éthanone
SMILES CC(=O)C1=C(SC(=C1)Cl)Cl
Spécification
Nom chimique ou matériau 3-Acetyl-2,5-dichlorothiophene
Point de fusion 37°C to 39°C
Point d’ébullition 120°C to 122°C (4 mmHg)
Point d’éclair 110°C (230°F)
Quantité 25 g
Beilstein 121288
Poids de la formule 195.07
Pourcentage de pureté 98%