Learn More
Thermo Scientific Alfa Aesar 50ML 2-Methyl-5-pyridylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal^t bott
Marque:  Thermo Scientific Alfa Aesar H58043.AD
50ML 2-Methyl-5-pyridylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal^t bott
Code nomenclature Nacres: NA.22
Informations supplémentaires : Poids : 0.00050kg Transport : UN number : 3399 Chem class : 4.3 Pack group : II
Code produit. 15415315