Learn More
Chlorure de 2-méthoxybenzoyle, 97 %, Thermo Scientific Chemicals
Code produit. 11309789
Click to view available options
Quantité:
5 g
25 g
100 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 21615-34-9
Formule moléculaire C8H7ClO2
Poids moléculaire (g/mol) 170.592
Numéro MDL MFCD00000664
Clé InChI RZNHSEZOLFEFGB-UHFFFAOYSA-N
Synonyme o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi
CID PubChem 88969
Nom IUPAC Chlorure de 2-méthoxybenzoyl
SMILES COC1=CC=CC=C1C(=O)Cl
Spécification
Nom chimique ou matériau 2-Methoxybenzoyl chloride
Densité 1.146
Point d’ébullition 128°C to 129°C (8 mmHg)
Point d’éclair 84°C (184°F)
Indice de réfraction 1.5724
Quantité 25 g
Numéro UN UN1729
Beilstein 607605
Sensibilité Moisture sensitive
Poids de la formule 170.6
Pourcentage de pureté 97%